Learn More
Medchemexpress LLC CTLA-4 Mouse HEK29 10ug
SDP
Catalog No. 5000196034
Encompass_Preferred
CTLA-4 Protein, Mouse (HEK293, His) is a recombinant extracellular domain of mouse CTLA-4 expressed in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. The product covers residues A37-D161, is formulated in phosphate buffer with physiological salt, and is intended for research use only. Follow the manufacturer's reconstitution and storage recommendations for best activity retention.
  • Expressed in HEK293 for mammalian post-translational modifications.
  • Includes a C-terminal 6xHis tag for purification and detection.
  • Covers residues A37-D161, representing the extracellular domain.
  • Lyophilized formulation for improved stability and transport.
  • Formulated in 20 mM PB, 150 mM NaCl, pH 7.4 for physiological compatibility.
  • Recommended handling: avoid reconstituting below 100 μg/mL and aliquot for long-term storage.
  • For research use only; not for diagnostic or therapeutic applications.
Catalog No. 50-001-96034 Supplier Medchemexpress LLC Supplier No. HYP7199110UG
CTLA-4 Protein, Mouse (HEK293, His) is a recombinant extracellular domain of mouse CTLA-4 expressed in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. The product covers residues A37-D161, is formulated in phosphate buffer with physiological salt, and is intended for research use only. Follow the manufacturer's reconstitution and storage recommendations for best activity retention.
  • Expressed in HEK293 for mammalian post-translational modifications.
  • Includes a C-terminal 6xHis tag for purification and detection.
  • Covers residues A37-D161, representing the extracellular domain.
  • Lyophilized formulation for improved stability and transport.
  • Formulated in 20 mM PB, 150 mM NaCl, pH 7.4 for physiological compatibility.
  • Recommended handling: avoid reconstituting below 100 μg/mL and aliquot for long-term storage.
  • For research use only; not for diagnostic or therapeutic applications.