Learn More
ACROBiosystems ACROBiosystems Human CTLA-4 / CD152 Protein, mouse IgG2a Fc Tag, low endotoxin
Catalog No. 501957483
Encompass_Preferred
Recombinant Protein;100UG;Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (CT4-H52A4) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession # NP_005205.2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 50 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and 100 kDa under non-reducing (NR) condition.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.1-2 ng/mL (QC tested).;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.5-7.8 ng/mL (Routinely tested).
Catalog No. 50-195-7483 Supplier ACROBiosystems Supplier No. CT4H52A4100UG
Recombinant Protein;100UG;Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (CT4-H52A4) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession # NP_005205.2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 50 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and 100 kDa under non-reducing (NR) condition.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.1-2 ng/mL (QC tested).;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 2 μg/mL (100 μL/well) can bind Human CTLA-4, mouse IgG2a Fc tag, low endotoxin (Cat. No. CT4-H52A4) with a linear range of 0.5-7.8 ng/mL (Routinely tested).