Learn More
Sino Biological Recombinant Human CTLA-4 Protein (His Tag), Amine-Labeled Biotinylated 100
Supplier:  Sino Biological 11159H08HB100
Encompass_Preferred
A DNA sequence encoding the human CTLA4 (NP_005205.2) extracellular domain (Met1-Phe162) was fused with the a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6169