Learn More
Medchemexpress LLC CTLA-4 Rat HEK293 5ug
SDP
Catalog No. 5000178509
Encompass_Preferred
CTLA-4 protein inhibits T-cell responses by binding strongly to CD80 and CD86 receptors surpassing CD28 This affinity allows CTLA-4 to effectively regulate T-cell activation and immune responses The balance between inhibitory and stimulatory signals controlled by CTLA-4 and its ligands modulates T-cell-mediated immune reactions CTLA-4 Protein Rat (HEK293 His) is the recombinant rat-derived CTLA-4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78509 Supplier Medchemexpress LLC Supplier No. HYP742155UG
Add to Cart
Add to Cart
CTLA-4 protein inhibits T-cell responses by binding strongly to CD80 and CD86 receptors surpassing CD28 This affinity allows CTLA-4 to effectively regulate T-cell activation and immune responses The balance between inhibitory and stimulatory signals controlled by CTLA-4 and its ligands modulates T-cell-mediated immune reactions CTLA-4 Protein Rat (HEK293 His) is the recombinant rat-derived CTLA-4 protein expressed by HEK293 with C-His labeled tag