Learn More
Medchemexpress LLC CTLA-4 Rhesus Macaq 5ug | 5ug
SDP
Catalog No. 5000187259
Encompass_Preferred
CTLA-4 protein acts as a primary inhibitory receptor playing a major role in regulating T-cell responses Its strong affinity for CD80 and CD86 receptors allows CTLA-4 to effectively suppress T-cell activation and maintain immune balance This interplay is crucial for fine-tuning T-cell-mediated immunity CTLA-4 Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived CTLA-4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87259 Supplier Medchemexpress LLC Supplier No. HYP773455UG
Add to Cart
Add to Cart
CTLA-4 protein acts as a primary inhibitory receptor playing a major role in regulating T-cell responses Its strong affinity for CD80 and CD86 receptors allows CTLA-4 to effectively suppress T-cell activation and maintain immune balance This interplay is crucial for fine-tuning T-cell-mediated immunity CTLA-4 Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived CTLA-4 protein expressed by HEK293 with C-His labeled tag