Learn More
Genetex Inc CTLA4 Monoclonal Antibody [MSVA-152R] HistoMAX™ (GTX04439) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04439
Encompass_Preferred
The CTLA4 antibody [MSVA-152R] HistoMAX™ (GTX04439) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human diffuse large B-cell lymphoma (DLBCL) tissue using GTX04439 CTLA4 antibody [MSVA-152R] HistoMAX. Diffuse large B cell lymphoma containing a large number of CTLA4 positive lymphocytes.
Catalog No. NC3666954