Learn More
Sino Biological CTLA-4 / CD152 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11159MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CTLA-4 / CD152 (rh CTLA-4 / CD152; Catalog#11159-H08H; NP_005205.2; Met1-Phe162). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8397