Learn More
Enzo Life Sciences CTLA4 monoclonal antibody (1E6) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6960100
Encompass_Preferred
Clone: 1E6. Isotype: IgG1. Immunogen: Extracellular domain of human CTLA-4. Species reactivity: Human. Applications: ICC, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3235