Learn More
ACROBiosystems Recombinant Protein;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag (Avitag™);HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501659184
Encompass_Preferred
Recombinant Protein;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag (Avitag™);200UG;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries an Avi tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 20-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-63 ng/mL (Routinely tested).
Catalog No. 50-165-9184 Supplier ACROBiosystems Supplier No. CT4C82E5200UG
Recombinant Protein;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag (Avitag™);200UG;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries an Avi tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 20-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avi Tag, His Tag (Cat. No. CT4-C82E5) with a linear range of 2-63 ng/mL (Routinely tested).