Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein, Fc Tag;HEK293;1MG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501659114
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (CT4-C5256) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40 kDa. As a result of glycosylation, the protein migrates as 45-52 kDa under reducing (R) condition, and 90-105 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-9114 Supplier ACROBiosystems Supplier No. CT4C52561MG
Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque CTLA-4, Fc Tag (CT4-C5256) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40 kDa. As a result of glycosylation, the protein migrates as 45-52 kDa under reducing (R) condition, and 90-105 kDa under non-reducing (NR) condition (SDS-PAGE).