Learn More
ACROBiosystems Recombinant Protein;Mouse CTLA-4 / CD152 Protein, Fc Tag;HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501658733
Encompass_Preferred
Recombinant Protein;Mouse CTLA-4 / CD152 Protein, Fc Tag;200UG;Mouse CTLA-4, Fc Tag (CT4-M5256) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession # NP_033973).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, His Tag (Cat. No. CD0-M5228) at 2μg/mL (100 μL/well) can bind Mouse CTLA-4, Fc Tag (Cat. No. CT4-M5256) with a linear range of 0.12-0.5 ng/mL (QC tested).
Catalog No. 50-165-8733 Supplier ACROBiosystems Supplier No. CT4M5256200UG
Recombinant Protein;Mouse CTLA-4 / CD152 Protein, Fc Tag;200UG;Mouse CTLA-4, Fc Tag (CT4-M5256) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession # NP_033973).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-1, His Tag (Cat. No. CD0-M5228) at 2μg/mL (100 μL/well) can bind Mouse CTLA-4, Fc Tag (Cat. No. CT4-M5256) with a linear range of 0.12-0.5 ng/mL (QC tested).