Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein;HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501659115
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein;200UG;Cynomolgus / Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.1 kDa. The protein migrates as 18-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque B7-1, Fc Tag (Cat. No. CD0-C5252) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Cynomolgus / Rhesus macaque B7-2, Fc Tag (Cat. No. CD6-C5254) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) with a linear range of 4-31 ng/mL (Routinely tested).
Catalog No. 50-165-9115 Supplier ACROBiosystems Supplier No. CT4C5227200UG
Recombinant Protein;Cynomolgus / Rhesus macaque CTLA-4 Protein;200UG;Cynomolgus / Rhesus macaque CTLA-4, His Tag (CT4-C5227) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.1 kDa. The protein migrates as 18-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque B7-1, Fc Tag (Cat. No. CD0-C5252) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Cynomolgus / Rhesus macaque B7-2, Fc Tag (Cat. No. CD6-C5254) at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CTLA-4, His Tag (Cat. No. CT4-C5227) with a linear range of 4-31 ng/mL (Routinely tested).