Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CTLA-4, His Tag, Avi Tag (Avitag™);HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Catalog No. 501658063
Encompass_Preferred
Recombinant Protein;Biotinylated Human CTLA-4, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CTLA-4, His Tag, Avi Tag (CT4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession # Q6GR94-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.2 kDa. The protein migrates as 22-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CTLA-4, His Tag, Avi Tag (Cat. No. CT4-H82E1) with a linear range of 0.8-25 ng/mL (QC tested).
Catalog No. 50-165-8063 Supplier ACROBiosystems Supplier No. CT4H82E1200UG
Recombinant Protein;Biotinylated Human CTLA-4, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CTLA-4, His Tag, Avi Tag (CT4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession # Q6GR94-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.2 kDa. The protein migrates as 22-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CTLA-4, His Tag, Avi Tag (Cat. No. CT4-H82E1) with a linear range of 0.8-25 ng/mL (QC tested).