Learn More
ATCC CTLL-2; T Lymphocyte; Mouse (Mus musculus)
Catalog No. 502383976
Encompass
CTLL-2 is a clone of cytotoxic T cells derived from a C57BL/6 mouse. The cells are dependent upon Interleukin-2 (IL-2) for growth and can be used to assay for IL-2.
Catalog No. 50-238-3976 Supplier ATCC Supplier No. TIB214
 Restricted Item
Add to Cart
Add to Cart
CTLL-2 is a clone of cytotoxic T cells derived from a C57BL/6 mouse. The cells are dependent upon Interleukin-2 (IL-2) for growth and can be used to assay for IL-2.