Learn More
MSC Tape Logic Packing Tape, 2 in, Red, Acrylic, 36 Rolls
Supplier: MSC 39581079
This colored carton sealing tape, made from thick polypropylene with acrylic adhesive, is ideal for color-coding shipments, dating inventory, and warehouse routing. It grips well to various surfaces, including recycled corrugated cardboard, ensuring a secure seal in heat, cold, and humid conditions. Application temperature: 32-140 degrees Fahrenheit.
- Thick pressure sensitive polypropylene tape with acrylic adhesive
- Grips well to a wide variety of surfaces, including recycled corrugated cardboard boxes
- Provides a secure seal under heat, cold and humid conditions
- Application temperature range is 32-140 degrees Fahrenheit
- Thickness of 2.2 mil
- Tensile strength of 21.4 lb/inch
- Color codes shipments or products
- Ideal for dating inventory and warehouse routing or location
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.