Learn More
MSC Tape Logic Packing Tape, 2 in, Red-White, 55 yd
Catalog No. 501918487
Encompass
This pre-printed carton sealing tape is designed to communicate special handling instructions while securely sealing cartons. It ensures important instructions stand out, aids in deterring pilferage, and alerts recipients of potential tampering.
  • Professional grade waterproof acrylic adhesive for a secure hold
  • Grips well to recycled corrugated boxes
  • Made from durable polypropylene material
  • Features a 2.2 mil thickness
  • Provides 21.4 lb/in tensile strength
  • Pre-printed in red and white for high visibility
Catalog No. 50-191-8487 Supplier MSC Supplier No. 39580774
This pre-printed carton sealing tape is designed to communicate special handling instructions while securely sealing cartons. It ensures important instructions stand out, aids in deterring pilferage, and alerts recipients of potential tampering.
  • Professional grade waterproof acrylic adhesive for a secure hold
  • Grips well to recycled corrugated boxes
  • Made from durable polypropylene material
  • Features a 2.2 mil thickness
  • Provides 21.4 lb/in tensile strength
  • Pre-printed in red and white for high visibility