Learn More
Genscript Corporation cTnI 6D11 mAb Mouse
Supplier:  Genscript Corporation V00302100
Encompass
Cardiac Troponin I (cTnI) is synthesized in cardiac muscle and forms a structural complex with Troponin T and C Its concentration in blood increases after the onset of chest pain and maintains peak for a couple of weeks The cTnI protein is considered as useful diagnosis marker for acute myocardial infarction (AMI)
Catalog No. 50-000-56789
Add to Cart
Request a Quote