Learn More
MyBioSource Mouse C-Peptide ELISA Kit
SDP
Supplier:  MyBioSource MBS76617596STRIPWELLS
Encompass_Preferred
The MyBioSource Mouse C-Peptide ELISA Kit quantifies C-Peptide in various mouse biological fluids using a 96-well plate coated with a specific antibody. Samples and standards are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and a TMB substrate that elicits a color change proportional to C-Peptide levels, measured at 450 nm.
Catalog No. NC1834002