Learn More
Genscript Corporation Cardiac Troponin I (cTnI), His, Human
Supplier:  Genscript Corporation Z0332050
Encompass
Cardiac Troponin I (cTnI) is a subtype of the troponin family that is commonly used as a marker for myocardial damage. Cardiac troponin I is specific for cardiac tissue and is detected in the serum only if myocardial injury has occurred. Because cardiac troponin I is a very sensitive and specific indicator of heart muscle (myocardium) damage, serum levels can be used to help differentiate between unstable angina and myocardial infarction (heart attack) in people with chest pain or acute coronary syndrome.
Catalog No. 50-227-2315