Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1499MSM8P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CTNNB1/1508. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa or MCF-7 cells. Breast carcinoma. Cellular Staining: Cell surface and cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length beta-Catenin (p120) protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 92kDa.Chromosome: 3p22.1. Gene Symbol: CTNNB1. Gene ID: 1499. Ensemble: ENSG00000168036. Uniprot: NP_001895.
Catalog No. 50-188-4633