Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1499RBP1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: Polyclonal. Clonality: Polyclonal. Host: Rabbit. Isotype: IgG. Positive Control: HeLa or MCF-7 cells. Breast carcinoma. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide from the middle of beta-Catenin (p120) protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 92kDa.Chromosome: 3p22.1. Gene Symbol: CTNNB1. Gene ID: 1499. Ensemble: ENSG00000168036. Uniprot: NP_001895.
Catalog No. 50-188-4635