Learn More
Aviva Systems Biology Corp CTNNB1 Antibody - C-terminal region (AVARP14001_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP14001_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CTNNB1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CTNNB1. Alternate names: CTNNBApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-53795