Learn More
Aviva Systems Biology Corp CTNNB1 Antibody - N-terminal region (P100600_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100600_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CTNNB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CTNNB1. Alternate names: CTNNBApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-62656