Learn More
Aviva Systems Biology Corp CTNND1 Antibody - middle region (ARP51890_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51890_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTNND1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CTNND1. Alternate names: CAS, CTNND, KIAA0384, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-604