Learn More
Apexbio Technology LLC CTOP TFA 1mg
SDP
Catalog No. 5000096028
Encompass_Preferred
CTOP TFA is a small-molecule antagonist targeting the -opioid receptor (MOR) It is designed to selectively inhibit MOR thereby regulating opioid receptor-mediated neural signaling pathways CTOP TFA exerts its biological activity primarily through specific interaction with and inhibition of MOR in the central nervous system Based on these pharmacological properties CTOP TFA holds research potential in investigating pain perception analgesia tolerance dependence neurotransmitter release stress responses and emotional regulation
Catalog No. 50-000-96028 Supplier Apexbio Technology LLC Supplier No. A89751
CTOP TFA is a small-molecule antagonist targeting the -opioid receptor (MOR) It is designed to selectively inhibit MOR thereby regulating opioid receptor-mediated neural signaling pathways CTOP TFA exerts its biological activity primarily through specific interaction with and inhibition of MOR in the central nervous system Based on these pharmacological properties CTOP TFA holds research potential in investigating pain perception analgesia tolerance dependence neurotransmitter release stress responses and emotional regulation