Learn More
Medchemexpress LLC HY-P1329A 1mg Medchemexpress, CTOP (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1329A1MG
Encompass_Preferred
Medchemexpress, HY-P1329A 1mg CTOP (TFA) CAS: CTOP TFA is a peptide that acts as a μ-opioid receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3050