Learn More
Cayman Chemical CTOP (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502940163
Encompass_Preferred
An antagonist of the μ-opioid receptor (IC50 = 2.8 nM); selective for μ-opioid receptors over δ-opioid receptors (IC50 = 13,500 nM) and somatostatin receptors (IC50 = 24,000 nM); inhibits the antinociceptive effect of morphine in the tail flick test in mice (ED50 = 0.018 nmol) and reverses morphine-induced increases in locomotor activity (ED50 = 0.02 nmol); induces withdrawal symptoms in a mouse model of chronic morphine dependence
Catalog No. 50-294-0163 Supplier Cayman Chemical Supplier No. 2737710 MG
An antagonist of the μ-opioid receptor (IC50 = 2.8 nM); selective for μ-opioid receptors over δ-opioid receptors (IC50 = 13,500 nM) and somatostatin receptors (IC50 = 24,000 nM); inhibits the antinociceptive effect of morphine in the tail flick test in mice (ED50 = 0.018 nmol) and reverses morphine-induced increases in locomotor activity (ED50 = 0.02 nmol); induces withdrawal symptoms in a mouse model of chronic morphine dependence