Learn More
Medchemexpress LLC CTP (Cardiac targeting peptide) | 1052692-86-0 | 99.7% | 1432.54 | 5 MG
SDP
Catalog No. 5000398473
Encompass_Preferred
CTP (cardiac targeting peptide) is a peptide that can transduce cardiomyocytes in vitro. It leads to efficient and specific transduction of heart tissue in mice models. CTP can be reversibly linked (e.g., via enolases, thiol groups) to cargo (e.g., miRNAs) for delivery specifically to cardiomyocytes over all other organs. For research use only.
  • Specific cardiomyocyte transduction in vitro and in mouse models.
  • Reversibly links to cargo like miRNAs for targeted delivery to cardiomyocytes.
  • Significant internalization in H9C2 cells when coupled with 6-CF.
  • Delivers peptide NBD into H9C2 cells, inhibiting TNF-α induced NF-κB transcriptional activity.
  • Delivers miRNA106a, reversing hypertrophic responses in human cardiomyocytes.
  • Rapidly transduces heart tissue in BALB/c mice in vivo.
  • High purity of 99.66% ensures reliable experimental results.
Catalog No. 50-003-98473 Supplier Medchemexpress LLC Supplier No. HYP40945MG
CTP (cardiac targeting peptide) is a peptide that can transduce cardiomyocytes in vitro. It leads to efficient and specific transduction of heart tissue in mice models. CTP can be reversibly linked (e.g., via enolases, thiol groups) to cargo (e.g., miRNAs) for delivery specifically to cardiomyocytes over all other organs. For research use only.
  • Specific cardiomyocyte transduction in vitro and in mouse models.
  • Reversibly links to cargo like miRNAs for targeted delivery to cardiomyocytes.
  • Significant internalization in H9C2 cells when coupled with 6-CF.
  • Delivers peptide NBD into H9C2 cells, inhibiting TNF-α induced NF-κB transcriptional activity.
  • Delivers miRNA106a, reversing hypertrophic responses in human cardiomyocytes.
  • Rapidly transduces heart tissue in BALB/c mice in vivo.
  • High purity of 99.66% ensures reliable experimental results.