Learn More
Medchemexpress LLC CTP inhibitor | 412940-35-3 | MFCD01122300 | 98.0% | 356.74 | C13H9ClN2O6S | 25 MG
SDP
Catalog No. 5000248047
Encompass_Preferred
CTP inhibitor (Compound 792949) is a research-grade small molecule that selectively inhibits the plasma membrane citrate transporter (PMCT/CTP). It is supplied as a high-purity solid for use in biochemical and cell-based studies of citrate transport and metabolism.
  • Potent and selective inhibitor of the plasma membrane citrate transporter.
  • High purity (98.0%) suitable for in vitro research.
  • Supplied as a solid; also available as 10 mM solution in DMSO.
  • Multiple size options, including 25 mg.
  • Storage: store at 4°C protected from light; in solvent, -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-48047 Supplier Medchemexpress LLC Supplier No. HY10199825MG
CTP inhibitor (Compound 792949) is a research-grade small molecule that selectively inhibits the plasma membrane citrate transporter (PMCT/CTP). It is supplied as a high-purity solid for use in biochemical and cell-based studies of citrate transport and metabolism.
  • Potent and selective inhibitor of the plasma membrane citrate transporter.
  • High purity (98.0%) suitable for in vitro research.
  • Supplied as a solid; also available as 10 mM solution in DMSO.
  • Multiple size options, including 25 mg.
  • Storage: store at 4°C protected from light; in solvent, -80°C (6 months), -20°C (1 month).