Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ctp inhibitor (compound 792949) | 412940-35-3 | MFCD00682824 | 98.0% | 356.74 g/mol | C13H9ClN2O6S | 50 MG
SDP

Catalog No. 5000248616
Encompass_Preferred

CTP inhibitor (Compound 792949) is a potent and selective inhibitor of the mitochondrial citrate transport protein (CTP). It reduces CTP activity while showing minimal inhibition of the plasma membrane citrate transporter (PMCT), and is supplied as an off-white to light yellow solid for research applications.

  • Potent and selective inhibition of mitochondrial citrate transport protein (CTP).
  • Minimal activity against plasma membrane citrate transporter (PMCT) at tested concentrations.
  • High purity (98.0%).
  • Molecular weight 356.74 g/mol.
  • Available in multiple sizes, including 50 MG and DMSO solution formats.
  • Off-white to light yellow solid appearance.
  • Storage recommendations: 4°C protected from light; in solvent -80°C (6 months), -20°C (1 month).
  • Intended for in vitro research use.

Catalog No. 50-002-48616 Supplier Medchemexpress LLC Supplier No. HY10199850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CTP inhibitor (Compound 792949) is a potent and selective inhibitor of the mitochondrial citrate transport protein (CTP). It reduces CTP activity while showing minimal inhibition of the plasma membrane citrate transporter (PMCT), and is supplied as an off-white to light yellow solid for research applications.

  • Potent and selective inhibition of mitochondrial citrate transport protein (CTP).
  • Minimal activity against plasma membrane citrate transporter (PMCT) at tested concentrations.
  • High purity (98.0%).
  • Molecular weight 356.74 g/mol.
  • Available in multiple sizes, including 50 MG and DMSO solution formats.
  • Off-white to light yellow solid appearance.
  • Storage recommendations: 4°C protected from light; in solvent -80°C (6 months), -20°C (1 month).
  • Intended for in vitro research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.