Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CTP Solution, Tris buffered

Catalog No. FERR1451 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1451 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1451 Supplier Thermo Scientific™ Supplier No. R1451
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ CTP (cytidine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base.

Thermo Scientific™ CTP (cytidine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base. The nucleotide has greater than 99% purity and is free of nuclease activities and functionally tested in in vitro transcription.

Features

  • Purity confirmed by HPLC
  • Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
  • Higher in vitro transcription yields using Tris buffered NTPs in comparison to NTPs titrated with NaOH
  • Stable for 3 years at -20°C and after multiple freeze-thaw cycles

Applications

  • in vitro transcription
  • RNA amplification
  • siRNA synthesis
  • aRNA synthesis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 100 mM CTP, Tris buffered (0.25 mL)

Store at –15 to –25°C.
Shipping Condition Dry Ice
Concentration 100 mM
Label or Dye Unlabeled
Quantity 0.25 mL
For Use With (Application) in vitro Transcription, RNA Amplification
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.