Learn More
Active Motif CTPB
SDP
Catalog No. NC1840795
Encompass
Active Motif's A selective activator of p300 HAT (histone acetyltransferase). Selective over PCAF (p300 CPB-associated factor).
Catalog No. NC1840795 Supplier Active Motif Supplier No. 14064
Active Motif's A selective activator of p300 HAT (histone acetyltransferase). Selective over PCAF (p300 CPB-associated factor).