Learn More
Enzo Life Sciences CTPB (5mg). CAS: 586976-24-1
SDP
Supplier:  Enzo Life Sciences ALX420033M005
Encompass_Preferred
Potent activator of p300 HAT (histone acetyltransferase), but not of PCAF (p300/CBP-associated factor) HAT activities. Alternative name: N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-6-pentadecyl-benzamide. Purity: ≥97% (TLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9111