Learn More
Medchemexpress LLC HY-124960 5mg Medchemexpress, CTPB CAS:586976-24-1 Purity:>98%
SDP
Catalog No. 502081062
Encompass_Preferred
Medchemexpress, HY-124960 5mg CTPB CAS:586976-24-1 CTPB is a good activator of p300 histone acetyl transferase (HAT) enzyme. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1062 Supplier Medchemexpress LLC Supplier No. HY1249605MG
Medchemexpress, HY-124960 5mg CTPB CAS:586976-24-1 CTPB is a good activator of p300 histone acetyl transferase (HAT) enzyme. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.