Learn More
AdipoGen CTPB
SDP
Catalog No. 502562361
Encompass_Preferred
Chemical. CAS 586976-24-1. Formula C31H43ClF3NO2. MW 554.1. Potent activator of p300 HAT histone acetyltransferase activity, which has an important role in regulation of gene expression. Does not show PCAF p300/CBP-associated factor HAT activities. Membrane-impermeable that requires a carrier to pass the membrane barrier of cells.
Catalog No. 50-256-2361 Supplier AdipoGen Supplier No. AGCR10141M001
Chemical. CAS 586976-24-1. Formula C31H43ClF3NO2. MW 554.1. Potent activator of p300 HAT histone acetyltransferase activity, which has an important role in regulation of gene expression. Does not show PCAF p300/CBP-associated factor HAT activities. Membrane-impermeable that requires a carrier to pass the membrane barrier of cells.