Learn More
Medchemexpress LLC HY-123986 250mg , CTPI-2 CAS:68003-38-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123986250MG
Encompass_Preferred
HY-123986 250mg CTPI-2 CAS: 68003-38-3 CTPI-2 is a third-generation mitochondrial citrate carrier SLC25A1 inhibitor with a KD of 3.5 μM. CTPI-2 inhibits glycolysis, PPARγ, and its downstream target the glucose transporter GLUT4. CTPI-2 halts salient alterations of NASH reverting steatosis, preventing the evolution to steatohepatitis, reducing inflammatory macrophage infiltration in the liver and adipose tissue, and starkly mitigating obesity induced by a high-fat diet. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9234