Learn More
MSC Intertape Polymer Group Reinforced Tape, Kraft, Water-Activated, 6.4 mil, 8/Case
Catalog No. 501918579
Encompass
This tape is designed for fast, permanent adhesion and superior strength. It offers excellent flexibility for sealing carton corners, edges, and irregular surfaces.
  • Extra strong fiberglass yarns
  • Patented starch-based adhesive
  • Water-activated, not affected by extreme hot or cold temperatures
  • Environmentally friendly, yarn can be removed during repulping for easy recycling
Catalog No. 50-191-8579 Supplier MSC Supplier No. 39581699
This tape is designed for fast, permanent adhesion and superior strength. It offers excellent flexibility for sealing carton corners, edges, and irregular surfaces.
  • Extra strong fiberglass yarns
  • Patented starch-based adhesive
  • Water-activated, not affected by extreme hot or cold temperatures
  • Environmentally friendly, yarn can be removed during repulping for easy recycling