Learn More
Aviva Systems Biology Corp CTR9 Antibody - C-terminal region (ARP64690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTR9. It was validated on Western Blot. . Alternate names: KIAA0155, SH2BP1, TSBP, p150, p150TSPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-288-336