Learn More
Aviva Systems Biology Corp CTRC Antibody - N-terminal region (ARP33873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTRC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CTRC. Alternate names: CLCR, ELA4ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55182