Learn More
Sino Biological CTRC / chymotrypsin C Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11456T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTRC / chymotrypsin C (rh CTRC / chymotrypsin C; Catalog#11456-H08H; Q99895; Met1-Leu268). CTRC / chymotrypsin C specific IgG was purified by Human CTRC / chymotrypsin C affinity chromatography.
Catalog No. 50-224-8627