Learn More
Sino Biological CTRC / chymotrypsin C Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11456R102100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CTRC / chymotrypsin C (rh CTRC / chymotrypsin C; Catalog#11456-H08H; Q99895; Met1-Leu268).
Catalog No. 50-224-7876