Learn More
Aviva Systems Biology Corp CTRL Antibody - N-terminal region (ARP33864_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33864_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTRL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CTRL. Alternate names: CTRL1, MGC70821ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-52931