Learn More
Diapharma Group, Inc. CTRL LVL 1 PLASMA
SDP
Catalog No. NC9184962
Encompass
APC Resistance Control Plasma Level 1, 5 x 1mL, Composition: Lyophilized citrated, stabilized human plasma prepared from pools of plasma collected from healthy donors, Appearance: lyophilized powder, Color: Yellowish, Odor: Odorless
Catalog No. NC9184962 Supplier Diapharma Group, Inc. Supplier No. C822650
APC Resistance Control Plasma Level 1, 5 x 1mL, Composition: Lyophilized citrated, stabilized human plasma prepared from pools of plasma collected from healthy donors, Appearance: lyophilized powder, Color: Yellowish, Odor: Odorless