Learn More
Diapharma Group, Inc. CTRL LVL 2 PLASMA
SDP
Catalog No. NC9184963
Encompass
APC Resistance Control Plasma Level 2, 5 x 1mL, Composition: Lyophilized citrated, stabilized human plasma prepared from pools of plasma collected from healthy donors, Its donors carry the Factor V:Q506 mutation, Color: Yellowish
Catalog No. NC9184963 Supplier Diapharma Group, Inc. Supplier No. C822668
APC Resistance Control Plasma Level 2, 5 x 1mL, Composition: Lyophilized citrated, stabilized human plasma prepared from pools of plasma collected from healthy donors, Its donors carry the Factor V:Q506 mutation, Color: Yellowish