Learn More
AdipoGen CTRP1 human pAb
SDP
Supplier:  AdipoGen AG25A0114C100
Encompass_Preferred
Polyclonal Antibody. Recognizes a N-terminal region of CTRP1 full-length protein. Detects bands of ~17kDa and ~36kDa by Western blot. This antibody is CTRP1-specific. Source Guinea pig. Applications ELISA, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. CTRP1 Complement C1q tumor necrosis factor-related protein 1 C1QTNF1 belongs to a highly conserved family of adiponectin paralogs. Serum levels of CTRP1 are significantly higher in obese/diabetic animal models and in hypertensive patients compared to normal controls. CTRP1 may be a new molecular link between obesity and hypertension.
Catalog No. 50-256-3207