Learn More
AdipoGen CTRP3 human pAb
SDP
Supplier:  AdipoGen AG25A0107C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the N-terminal half of CTRP3 full-length protein. Detects bands of ~20kDa and ~35kDa by Western blot. Source Guinea pig. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. CTRP3 plays an important role in regulating both embryonic cartilage development and postnatal longitudinal bone growth. CTRP3 stimulation leads to ERK1/2-dependent cell proliferation in human osteosarcoma cell lines.
Catalog No. 50-256-3213