Learn More
AdipoGen CTRP5 Glob-Dom h pAb
SDP
Supplier:  AdipoGen AG25A0096C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human CTRP5 globular domain and CTRP5 full-length protein. Detects a band of ~18kDa and ~30kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. CTRP5 C1qTNF-related protein 5 C1QTNF5 belongs to a highly conserved family of adiponectin paralogs. CTRP5 mediates activation of AMP-activated protein kinase AMPK in muscle and liver cells, thereby regulating glucose and lipid metabolism. Serum levels of CTRP5 are significantly higher in obese/diabetic animal models compared to normal controls. Furthermore, CTRP5 may be a putative biomarker for mitochondrial dysfunction. Defects in C1QTNF5 are a cause of late-onset retinal degeneration LORD.
Catalog No. 50-256-3224