Learn More
AdipoGen CTRP5 human rec His
SDP
Catalog No. 502562977
Encompass_Preferred
Protein. Human CTRP5 aa 16-243 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Lyophilized from a 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. CTRP5 C1qTNF-related protein 5 C1QTNF5 belongs to a highly conserved family of adiponectin paralogs. CTRP5 mediates activation of AMP-activated protein kinase AMPK in muscle and liver cells, thereby regulating glucose and lipid metabolism. Serum levels of CTRP5 are significantly higher in obese/diabetic animal models compared to normal controls. Furthermore, CTRP5 may be a putative biomarker for mitochondrial dysfunction. Defects in C1QTNF5 are a cause of late-onset retinal degeneration LORD.
Catalog No. 50-256-2977 Supplier AdipoGen Supplier No. AG40A0142C050
Protein. Human CTRP5 aa 16-243 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Lyophilized from a 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl. Purity >90% SDS-PAGE. CTRP5 C1qTNF-related protein 5 C1QTNF5 belongs to a highly conserved family of adiponectin paralogs. CTRP5 mediates activation of AMP-activated protein kinase AMPK in muscle and liver cells, thereby regulating glucose and lipid metabolism. Serum levels of CTRP5 are significantly higher in obese/diabetic animal models compared to normal controls. Furthermore, CTRP5 may be a putative biomarker for mitochondrial dysfunction. Defects in C1QTNF5 are a cause of late-onset retinal degeneration LORD.