Learn More
AdipoGen anti-CTRP6 (human), mAb (256-E) (50 µg)
SDP
Supplier:  AdipoGen AG20A0093C050
Encompass_Preferred
Monoclonal Antibody. Recognizes the N-terminal half of human CTRP6 full-length protein. Detects bands of ~16kDa and ~32kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: 256-E. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. CTRP6 mediates the phosphorylation of AMPK and acetyl-CoA carboxylase (ACC) in skeletal muscle cells, subsequently mediating glycogen accumulation and fatty acid oxidation. It induces the expression of the potent anti-inflammatory cytokine IL-10 in macrophages. CTRP6 may be a potential serum marker for hepatocellular carcinoma, contributing to tumor angiogenesis by activating the Akt pathway. CTRP6 was also shown to be associated with obesity, type 1 diabetes mellitus (T1D), vitiligo (depigmentation of the skin) and ASFV (African Swine Fever Virus). CTRP6 forms homotrimers and higher-order oligomers and it may also form heteromeric complexes with CTRP1. May be involved in kidney inflammation.
Catalog No. 50-205-2896