Learn More
AdipoGen CTRP6 human pAb
SDP
Supplier:  AdipoGen AG25A0108C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the N-terminal half of CTRP6 full-length protein. Detects bands of ~16kDa and ~32kDa by Western blot. Source Guinea pig. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. CTRP6 belongs to a highly conserved family of adiponectin paralogs. CTRP6 mediates activation of AMP-activated protein kinase AMPK in skeletal muscle cells, thereby regulating glucose and lipid metabolism. Mediates the phosphorylation of acetyl-CoA carboxylase ACC.
Catalog No. 50-256-3212